Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethanol, ReagentPlus™, ≥99.0%, Solstice

Catalog No. 60045058 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Stainless Steel Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-058 20 L Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-045-058 Supplier Solstice Supplier No. 18546920L

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, contains 50 ppm BHT as stabilizer,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Melting Point -85°C
Color Colorless
Density 0.965g/mL at 25°C
pH 4 to 7
Boiling Point 124°C
Explosion Limit 0.245
Flash Point 40°C (104°F)
Ignition Point 548°F
Packaging Stainless Steel Can
Refractive Index n20/D 1.402(lit.)
Linear Formula CH3OCH2CH2OH
Quantity 20 L
UN Number UN1188
Beilstein 1731074
Vapor Pressure 6.17mmHg (20°C)
Formula Weight 76.09g/mol
Percent Purity ≥99.0%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
Hazard Statement P201-P210-P260-P280-P308 + P311-P370 + P378
Precautionary Statement H226-H302 + H312 + H332-H360FD-H370-H373
DOTInformation Class 3 - PG 3 - Ethylene glycol monomethyl ether
EINECSNumber 203-713-7
RTECSNumber KL5775000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.