Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethyl Acetate 98%, Thermo Scientific™

Catalog No. AC149350010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC149350010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC149350010 Supplier Thermo Scientific Chemicals Supplier No. 149350010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-49-6
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00008719
InChI Key XLLIQLLCWZCATF-UHFFFAOYSA-N
Synonym methyl glycol acetate, mecsac, acetic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, acetate, methylglykolacetat, ethylene glycol monomethyl ether acetate, methyl cellosolve acetate, acetyl methyl cellosolve, 2-methoxyethanol acetate, methyl glycol monoacetate
PubChem CID 8054
IUPAC Name 2-methoxyethyl acetate
SMILES COCCOC(C)=O

Specifications

Acidity 0.02% max.
CAS Min % 97.5
CAS Max % 100.0
Density 1.004
Infrared Spectrum Authentic
Refractive Index 1.4010 to 1.4030 (20°C, 589nm)
Linear Formula CH3CO2CH2CH2OCH3
Quantity 1 L
Beilstein 02, 141
Specific Gravity 1.001 to 1.005 (20°C)
Merck Index 15, 6064
Formula Weight 118.13
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl acetate, 98%
Show More Show Less
Hazard Category POSSIBLE TERATOGEN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.