Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxymethyl-3-methoxypropenenitrile, Mixture of cis and trans 97%, Thermo Scientific™

Catalog No. AC269620250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC269620250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC269620250 Supplier Thermo Scientific Chemicals Supplier No. 269620250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1608-82-8
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
InChI Key QZFRWRSOUHEOFM-GQCTYLIASA-N
Synonym 3-methoxy-2-methoxymethyl acrylonitrile, 2-methoxymethylene-3-methoxypropanenitrile, e-3-methoxy-2-methoxymethyl prop-2-enenitrile, 2e-3-methoxy-2-methoxymethyl prop-2-enenitrile, e-3-methoxy-2-methoxymethyl-2-propenenitrile
PubChem CID 2724688
IUPAC Name (E)-3-methoxy-2-(methoxymethyl)prop-2-enenitrile
SMILES COCC(=COC)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Infrared Spectrum Authentic
Refractive Index 1.4635 to 1.4655 (20°C, 589nm)
Linear Formula CH3OCH2C(CN)=CHOCH3
Quantity 25 mL
Formula Weight 127.15
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 2-Methoxymethyl-3-methoxypropenenitrile, mixture of cis and trans, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.