Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxynicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. M204125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M204125G 25 g
M20415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M204125G Supplier TCI America Supplier No. M204125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16498-81-0
Molecular Formula C7H6NO3
Molecular Weight (g/mol) 152.13
MDL Number MFCD00085110
InChI Key FTEZJSXSARPZHJ-UHFFFAOYSA-M
Synonym 2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh
PubChem CID 708093
IUPAC Name 2-methoxypyridine-3-carboxylate
SMILES COC1=NC=CC=C1C([O-])=O

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 25 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxynicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.