Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. I051225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I051225G 25 g
I05125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I051225G Supplier TCI America Supplier No. I051225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3288-04-8
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00011675
InChI Key QKAOOWJWWKWWOZ-UHFFFAOYSA-N
Synonym 2-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-2-methoxy, o-methoxyphenyl isothiocyanate, 2-methoxyphenylisothiocyanate, 1-isothiocyanato-2-methoxy-benzene, isothiocyanic acid 2-methoxyphenyl ester, 2-methoxybenzenisothiocyanate, acmc-1coqu, o-methoxyphenylisothiocyanat, orthomethoxyphenyl isothiocyanate
PubChem CID 520599
IUPAC Name 1-isothiocyanato-2-methoxybenzene
SMILES COC1=CC=CC=C1N=C=S

Specifications

Color Yellow
Boiling Point 267°C
Quantity 25 g
UN Number 1760
Formula Weight 165.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyphenyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.