Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. M27925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27925G 5 g
M27921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27925G Supplier TCI America Supplier No. M27925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59099-58-0
Molecular Formula C8H7F3O4S
Molecular Weight (g/mol) 256.20
MDL Number MFCD25541795
InChI Key RCDZTJFBROIDKD-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 2-Methoxyphenyl Ester, 2-Methoxyphenyl Triflate
PubChem CID 11780115
IUPAC Name 2-methoxyphenyl trifluoromethanesulfonate
SMILES COC1=CC=CC=C1OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 116°C
Quantity 5 g
Formula Weight 256.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyphenyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.