Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxypyridine-4-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M27305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2730-5G 5 g
M27301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2730-5G Supplier TCI America Supplier No. M27305G
Add to Cart
Add to Cart

Specifications

CAS 762262-09-9
Molecular Formula C6H8BNO3
MDL Number MFCD07368877
Quantity 5 g
Synonym 2-methoxypyridine-4-boronic acid, 2-methoxypyridin-4-yl boronic acid, 2-methoxypyridne-4-boronic acid, 2-methoxy-4-pyridineboronic acid, 2-methoxypyridin-4-yl-4-boronic acid
InChI Key DHQMUJSACXTPEA-UHFFFAOYSA-N
SMILES B(C1=CC(=NC=C1)OC)(O)O
IUPAC Name (2-methoxypyridin-4-yl)boronic acid
Molecular Weight (g/mol) 152.944
PubChem CID 23546919
Formula Weight 152.94
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.