Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxypyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M20291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20291G 1 g
M2029-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20291G Supplier TCI America Supplier No. M20291G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 2-Methoxypyridine-5-boronic Acid (contains varying amounts of Anhydride)
CAS 163105-89-3
Melting Point 138°C
Color White-Yellow
Molecular Formula C6H8BNO3
Quantity 1 g
MDL Number MFCD02093044
Synonym 2-methoxy-5-pyridineboronic acid, 6-methoxypyridin-3-yl boronic acid, 2-methoxypyridine-5-boronic acid, 2-methoxy-5-pyridinylboronic acid, 6-methoxypyridine-3-boronic acid, 6-methoxypyridin-3-yl-3-boronic acid, 6-methoxy-pyridine-3-boronic acid, 6-methoxy-3-pyridinylboronic acid, 6-methoxy-3-pyridylboronic acid, 6-methoxypyridin-3-yl boranediol
InChI Key DHADXDMPEUWEAS-UHFFFAOYSA-N
SMILES COC1=CC=C(C=N1)B(O)O
IUPAC Name (6-methoxypyridin-3-yl)boronic acid
Molecular Weight (g/mol) 152.94
PubChem CID 2734368
Formula Weight 152.94
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.