Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,3-cyclohexanedione 98.0+%, TCI America™
SDP

Catalog No. M113325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M113325G 25 g
M11335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M113325G Supplier TCI America Supplier No. M113325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1193-55-1
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00001587
InChI Key VSGJHHIAMHUZKF-UHFFFAOYSA-N
Synonym 2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g
PubChem CID 70945
IUPAC Name 2-methylcyclohexane-1,3-dione
SMILES CC1C(=O)CCCC1=O

Specifications

Quantity 25 g
Formula Weight 126.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1,3-cyclohexanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.