Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methyl-1,3-dioxolane

Catalog No. 29220625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-220-625G 25 g
29-220-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-220-625G Supplier Sigma Aldrich Supplier No. 29220625G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The kinetics and mechanism of the gas-phase thermal decomposition of 2-methyl-1,3-dioxolane has been studied in a static system
  • The infrared spectra of solid, liquid and gaseous 2-methyl-1,3-dioxolane has been studied
  • Low-temperature ozonation of 2-methyl-1,3-dioxolane in acetone-d6, methyl acetate and tert-butyl methyl ether has been reported

Specifications

CAS 497-26-7
Density 0.982 g/mL (at 25°C (literature))
Boiling Point 82°C to 83°C (lit.)
Molecular Formula C4H8O2
Refractive Index n20/D 1.398 (literature)
Linear Formula C4H8O2
MDL Number MFCD00003210
Quantity 25 g
Synonym Acetaldehyde ethylene acetal
Molecular Weight (g/mol) 88.11
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.