Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,3-propanediamine 98.0+%, TCI America™
SDP

Catalog No. M16571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16571G 1 g
M16575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16571G Supplier TCI America Supplier No. M16571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2400-78-4
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.15
MDL Number MFCD08276399
InChI Key BDXGMDGYOIWKIF-UHFFFAOYSA-N
Synonym 1,3-Diamino-2-methylpropane
PubChem CID 14499052
IUPAC Name 2-methylpropane-1,3-diamine
SMILES CC(CN)CN

Specifications

Color Colorless
Quantity 1 g
UN Number 2734
Formula Weight 88.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1,3-propanediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.