Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 2-Methyl-1,3-propanediol, 98+%

Catalog No. AAH36922A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH36922A1 1 kg
AAH3692230 250 g
AAH36922A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH36922A1 Supplier Thermo Scientific™ Supplier No. H36922A1
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2163-42-0 | C4H10O2 | 90.12 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2163-42-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00082586
InChI Key QWGRWMMWNDWRQN-UHFFFAOYSA-N
Synonym Methylpropanediol;
IUPAC Name 2-methylpropane-1,3-diol
SMILES CC(CO)CO

Specifications

Melting Point -54°
Density 1.015
Boiling Point 123-125/20mm
Flash Point 127°(261°F)
Quantity 1 kg
Formula Weight 90.12
Percent Purity 98+%
Physical Form Liquid
ShelfLife 5 Years
EINECSNumber 606-809-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.