Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,3-propanediol 98.0+%, TCI America™
SDP

Catalog No. M111425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M111425ML 25 mL
M1114500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M111425ML Supplier TCI America Supplier No. M111425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2163-42-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00082586
InChI Key QWGRWMMWNDWRQN-UHFFFAOYSA-N
Synonym 1,3-Dihydroxy-2-methylpropane
PubChem CID 75103
IUPAC Name 2-methylpropane-1,3-diol
SMILES CC(CO)CO

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 mL
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1,3-propanediol
EINECSNumber (2)-3203
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.