Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,4-naphthoquinone, 98%

Catalog No. AAA1359318
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1359318 50 g
AAA1359309 10 g
AAA1359330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1359318 Supplier Thermo Scientific Chemicals Supplier No. A1359318
Add to Cart

CAS: 58-27-5 | C11H8O2 | 172.183 g/mol

2-Methyl-1,4-naphthoquinone is a prothrombogenic compound and is used as a model quinone in cell culture investigations. 2-Methyl-1,4-naphthoquinone is a synthetic analogue of of 1,4-naphthoquinone with a methyl group in the 2-position. 2-Methyl-1,4-naphthoquinone can be used as an oxidative injury inducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1,4-naphthoquinone is a prothrombogenic compound and is used as a model quinone in cell culture investigations. 2-Methyl-1,4-naphthoquinone is a synthetic analogue of of 1,4-naphthoquinone with a methyl group in the 2-position. 2-Methyl-1,4-naphthoquinone can be used as an oxidative injury inducing agent.

Solubility
Soluble in methanol

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-27-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00001681
InChI Key MJVAVZPDRWSRRC-UHFFFAOYSA-N
Synonym menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
PubChem CID 4055
ChEBI CHEBI:28869
IUPAC Name 2-methylnaphthalene-1,4-dione
SMILES CC1=CC(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 104°C to 107°C
Odor Odorless
Quantity 50 g
Beilstein 1908453
Sensitivity Light sensitive
Merck Index 14,5831
Solubility Information Soluble in methanol
Formula Weight 172.18
Percent Purity 98%
Chemical Name or Material 2-Methyl-1,4-naphthoquinone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-372-6
RTECSNumber QL9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.