Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methyl-1-butene

Catalog No. 502891503
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-1503 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-289-1503 Supplier Sigma Aldrich Supplier No. 25748625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methyl-1-butene
TRUSTED_SUSTAINABILITY

Specifications

CAS 563-46-2
Melting Point -137°C (lit.)
Density 0.65 g/mL (at 25°C (literature))
Boiling Point 31°C (lit.)
Molecular Formula C5H10
Refractive Index n20/D 1.378 (literature)
Linear Formula CH3CH2C(CH3)=CH2
MDL Number MFCD00009333
Quantity 25 g
Molecular Weight (g/mol) 70.13
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.