Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-Propanol, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99% (GC), Solstice

Catalog No. 60046181 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-181 1 L Poly Bottle
60-046-182 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-181 Supplier Solstice Supplier No. 330641L
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point -108°C
Color Colorless
Density 0.803g/mL at 25°C
pH 7 (at 80g/L)
Boiling Point 108°C
Flash Point 28°C (82.4°F)
Ignition Point 801°F
Packaging Poly Bottle
Refractive Index n20/D 1.396
Linear Formula (CH3)2CHCH2OH
Quantity 1 L
UN Number UN1212
Beilstein 1730878
Solubility Information In water 70g/L at 20°C
Vapor Density 2.55 (vs air)
Vapor Pressure 8mmHg (20°C), 8.8mmHg (0°C)
Formula Weight 74.12g/mol
Percent Purity ≥99% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material 2-Methyl-1-Propanol, Reag. Ph. Eur.
Show More Show Less
Hazard Statement P210-P280-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
Precautionary Statement H226-H315-H318-H335-H336
DOTInformation Class 3 - PG 3 - Isobutanol
EINECSNumber 201-148-0
RTECSNumber NP9625000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.