Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-pyrroline 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
50mL
Packaging:
Glass bottle
872-32-2
C5H9N
83.13
MFCD00128806
CTSZPNIMMLSKDV-UHFFFAOYSA-N
2-methyl-1-pyrroline,3,4-dihydro-5-methyl-2h-pyrrole,1pyrroline2methyl,2h-pyrrole, 3,4-dihydro-5-methyl,pubchem9412
70103
CHEBI:78856
5-methyl-3,4-dihydro-2H-pyrrole
CC1=NCCC1
872-32-2
C5H9N
83.13
MFCD00128806
CTSZPNIMMLSKDV-UHFFFAOYSA-N
2-methyl-1-pyrroline,3,4-dihydro-5-methyl-2h-pyrrole,1pyrroline2methyl,2h-pyrrole, 3,4-dihydro-5-methyl,pubchem9412
70103
CHEBI:78856
5-methyl-3,4-dihydro-2H-pyrrole
CC1=NCCC1
Specific Gravity 0.878
Boiling Point 104.0°C to 105.0°C
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Water 2% max.
Flash Point 11°C
Density 0.8780g/mL
Quantity 25mL
Infrared Spectrum Authentic
Color Beige to White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: soluble.
Formula Weight 83.13
Physical Form Crystalline Powder and/or Chunks
Percent Purity 98%
Chemical Name or Material 2-Methyl-1-pyrroline, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-822-9

TSCA : TSCA