Learn More
Sigma Organic Chemistry 2-Methyl-2-butene | 100ML | 513-35-9 | MFCD00009276
Supplier: Sigma Organic Chemistry M32704100ML

2-Methyl-2-butene, 100ML
About This Item:
Synonym(s): beta-Isoamylene; Amylene
Linear Formula: CH3CH=C(CH3)2
Molecular Weight: 70.13
CAS Number: 513-35-9
MDL Number: MFCD00009276
UNSPSC Code: 12352100
Purity: >=99%
Boiling Point: 35-38 C (lit.)
Melting Point: -134 C (lit.)
Density: 0.662 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.385 (lit.)
Storage: room temp
EINECS Number: 208-156-3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.