Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-butene, 99+%

Catalog No. p-160658
Change view
Click to view available options
Quantity:
25 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126490250 25 mL Glass bottle
AC126492500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC126490250 Supplier Thermo Scientific Chemicals Supplier No. 126490250
Add to Cart
This item is not returnable. View return policy

2-Methyl-2-butene, 99+%, C5H10, CAS Number-513-35-9, 1,1,2-trimethylethylene, 2-butene, 2-methyl, 2-methyl-2-butene, 2-methylbutene-2, 3-methyl-2-butene, amylene, beta-isoamylene, ethylene, trimethyl, n-amylene, trimethylethylene, 250mL, 99% min. (GC), 01, 211, 35.0 deg.C to 38.0 deg.C, 100.0, 99.0 | CAS: 513-35-9 | C5H10 | 70.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-35-9
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009276
InChI Key BKOOMYPCSUNDGP-UHFFFAOYSA-N
Synonym 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
PubChem CID 10553
ChEBI CHEBI:77916
IUPAC Name 2-methylbut-2-ene
SMILES CC=C(C)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -134.0°C
Color Colorless to Yellow
Density 0.6600g/mL
Boiling Point 35.0°C to 38.0°C
Flash Point −45°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=C(CH3)2
Refractive Index 1.3860 to 1.3880
Quantity 25 mL
Beilstein 01, 211
Specific Gravity 0.66
Merck Index 15, 601
Formula Weight 70.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-butene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected o
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a well-ventilated place.
Keep cool.
Keep away from heat/sparks/open flame
EINECSNumber 208-156-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.