Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-furanthiol 95.0+%, TCI America™
SDP

Catalog No. M18475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18475G 5 g
M18471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18475G Supplier TCI America Supplier No. M18475G

Chemical Identifiers

CAS 28588-74-1
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00010280
InChI Key RUYNUXHHUVUINQ-UHFFFAOYSA-N
Synonym 2-methyl-3-furanthiol, 3-furanthiol, 2-methyl, 2-methyl-3-mercaptofuran, 2-methyl-3-furanethiol, 2-methyl-3-furylthiol, 2-methyl-furan-3-thiol, 3-mercapto-2-methylfuran, 2-methyl-3-furylmercaptan, 2-methyl-3-sulfanylfuran, unii-n21rw1n179
PubChem CID 34286
IUPAC Name 2-methylfuran-3-thiol
SMILES CC1=C(S)C=CO1

Specifications

Boiling Point 60°C
Quantity 5 g
UN Number 1228
Formula Weight 114.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-furanthiol
RTECSNumber LU6235000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.