Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
2-Methyl-3-nitroaniline
Spécifications
Spécifications
| CAS | 603-83-8 |
| Melting Point | 88°C to 90°C (lit.) |
| Boiling Point | 305°C (lit.) |
| Molecular Formula | C7H8N2O2 |
| Linear Formula | CH3C6H3(NO2)NH2 |
| MDL Number | MFCD00007731 |
| Quantity | 1 g |
| Synonym | 2-Amino-6-nitrotoluene; 3-Nitro-o-toluidine |
| Molecular Weight (g/mol) | 152.15 |
| Percent Purity | 97% |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'