Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-nitrobenzyl chloride, 97%, Thermo Scientific™

Catalog No. AC187400050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC187400050 5 g
AC187400010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC187400050 Supplier Thermo Scientific Chemicals Supplier No. 187400050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60468-54-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD00007165
InChI Key XZNDXQGZPOZITR-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrobenzyl chloride, 1-chloromethyl-2-methyl-3-nitrobenzene, alpha-chloro-2-methyl-3-nitrotoluene, 1-chloromethyl-2-methyl-3-nitro-benzene, 2-methyl-3-nitrobenzylchloride, benzene, 1-chloromethyl-2-methyl-3-nitro, acmc-1b2j5, ksc497o8t, 2-chloromethyl-6-nitrotoluene
PubChem CID 108947
IUPAC Name 1-(chloromethyl)-2-methyl-3-nitrobenzene
SMILES CC1=C(CCl)C=CC=C1[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 42°C to 47°C
Flash Point >109°C
Assay Percent Range 96% min. (GC)
Linear Formula CH3C6H3(NO2)CH2Cl
Quantity 5 g
Formula Weight 185.61
Percent Purity 97%
Chemical Name or Material 2-Methyl-3-nitrobenzyl chloride, 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 262-249-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.