Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methyl-3-pentanone

Catalog No. 10870725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-870-725G 25 g
10-870-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-870-725G Supplier Sigma Aldrich Supplier No. 10870725G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methyl-3-pentanone may be used to study the stereochemistry of ketone enolization by lithium 2, 2, 6, 6-tetramethylpiperidide.

Specifications

CAS 565-69-5
Density 0.811 g/mL (at 25°C (literature))
Boiling Point 113°C (lit.)
Molecular Formula C6H12O
Refractive Index n20/D 1.397 (literature)
Linear Formula (CH3)2 CHCOC2H5
MDL Number MFCD00009314
Quantity 25 g
Synonym Ethyl isopropyl ketone
Molecular Weight (g/mol) 100.16
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.