Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-(propyldithio)furan 97.0+%, TCI America™
SDP

Catalog No. M18401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18401G 1 g
M18405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18401G Supplier TCI America Supplier No. M18401G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61197-09-9
Molecular Formula C8H12OS2
Molecular Weight (g/mol) 188.303
MDL Number MFCD01729816
InChI Key YFPPCUTVJGGSQC-UHFFFAOYSA-N
Synonym 2-Methyl-3-furyl Propyl Disulfide
PubChem CID 43579
IUPAC Name 2-methyl-3-(propyldisulfanyl)furan
SMILES CCCSSC1=C(OC=C1)C

Specifications

Quantity 1 g
UN Number 2810
Formula Weight 188.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-(propyldithio)furan
RTECSNumber JO1975500
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.