Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-5-nitrobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. M257025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M257025G 25 g
M25705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M257025G Supplier TCI America Supplier No. M257025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 939-83-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00017040
InChI Key XOSDYLFXPMFRGF-UHFFFAOYSA-N
Synonym benzonitrile, 2-methyl-5-nitro, 5-nitro-o-tolunitrile, 2-cyano-4-nitrotoluene, 2-methyl-5-nitrobenzenecarbonitrile, 3-cyano-4-methylnitrobenzene, 2-methyl-5-nitro-benzonitrile, 4-nitro-2-cyanotoluene, 5-nitro-ortho-tolunitrile, 5-nitro-2-methylbenzonitril, 6-methyl-3-nitrobenzonitrile
PubChem CID 350143
IUPAC Name 2-methyl-5-nitrobenzonitrile
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 106°C
Color Yellow
Boiling Point 175°C
Quantity 25 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-5-nitrobenzonitrile
EINECSNumber (3)-1780
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.