Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-6-phenylpyridine 98.0+%, TCI America™
SDP

Catalog No. M20011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20011G 1 g
M20015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20011G Supplier TCI America Supplier No. M20011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 46181-30-0
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00796537
InChI Key GREMYQDDZRJQEG-UHFFFAOYSA-N
PubChem CID 2762885
IUPAC Name 2-methyl-6-phenylpyridine
SMILES CC1=CC=CC(=N1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 155°C
Quantity 1 g
Formula Weight 169.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methyl-6-phenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.