Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-8-nitroquinoline 98.0+%, TCI America™
SDP

Catalog No. N053925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N053925G 25 g
N05395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N053925G Supplier TCI America Supplier No. N053925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 881-07-2
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00006764
InChI Key UHPGVDHXHDPYQP-UHFFFAOYSA-N
PubChem CID 13433
IUPAC Name 2-methyl-8-nitroquinoline
SMILES CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O

Specifications

Melting Point 138°C
Color Yellow
Quantity 25 g
Synonym 8-nitroquinaldine, quinaldine, 8-nitro, quinoline, 2-methyl-8-nitro, unii-xrv949auqz, xrv949auqz, acmc-209qr6, 2-methyl-8-nitro-quinoline, 5-20-07-00387 beilstein handbook reference, quinoline,2-methyl-8-nitro, uhpgvdhxhdpyqp-uhfffaoysa
Formula Weight 188.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-8-nitroquinoline
RTECSNumber UZ9830000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.