Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzo[b]thiophene, 98%

Catalog No. AAH3222403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3222403 1 g
AAH3222406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3222403 Supplier Thermo Scientific Chemicals Supplier No. H3222403
Add to Cart

CAS: 1195-14-8 | C9H8S | 148.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195-14-8
Molecular Formula C9H8S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00216250
InChI Key BLZKSRBAQDZAIX-UHFFFAOYSA-N
PubChem CID 70952
IUPAC Name 2-methyl-1-benzothiophene
SMILES CC1=CC2=CC=CC=C2S1

Specifications

Melting Point 51°C to 52°C
Quantity 1 g
Synonym 2-methylbenzo b thiophene, 2-methylthianaphthene, 2-methyl-1-thiaindene, 2-methylbenzothiophene, 2-methylthioindene, benzo b thiophene, 2-methyl, benzo b thiophene,2-methyl, acmc-1bscn, benzothiophene, 2-methyl
Formula Weight 148.23
Percent Purity 98%
Chemical Name or Material 2-Methylbenzo[b]thiophene
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 214-792-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.