Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzothiazole, 99%

Catalog No. AC150380250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150380250 25 g Glass bottle
AC150381000 100 g Glass bottle
AC150385000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC150380250 Supplier Thermo Scientific Chemicals Supplier No. 150380250
Add to Cart

CAS: 120-75-2 | C8H7NS | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-75-2
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
InChI Key DXYYSGDWQCSKKO-UHFFFAOYSA-N
Synonym 2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w
PubChem CID 8446
IUPAC Name 2-methyl-1,3-benzothiazole
SMILES CC1=NC2=CC=CC=C2S1

Specifications

CAS Min % 98.5
CAS Max % 99.0
Melting Point 12.0°C to 14.0°C
Color Amber to Colorless
Density 1.2000g/mL
Boiling Point 238.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6150 to 1.6180
Quantity 25 g
Beilstein 27,46
Specific Gravity 1.2
Solubility Information Solubility in water: insoluble. Other solubilities: ssluble in ether and chloroform
Formula Weight 149.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylbenzothiazole, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 204-423-3
RTECSNumber DL5600000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.