Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbutane, Solstice

Catalog No. 60090052 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-052 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-052 Supplier Solstice Supplier No. M326316X1L/DEL

missing translation for 'validMainframeMsg'

Reagent Grade, ≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-78-4
PubChem CID 6556
ChEBI CHEBI:30362

Specifications

Quantity 6 x 1 L
Flash Point < -35 °C
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 28°C (1.013 hPa)
Percent Purity min. 99.0 %
Grade Reagent
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.