Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylcinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. M129525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M129525G 25 g
M12955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M129525G Supplier TCI America Supplier No. M129525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2373-76-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00016841
InChI Key RSWBWHPZXKLUEX-VOTSOKGWSA-N
Synonym 2-methylcinnamic acid, 3-o-tolyl acrylic acid, o-methylcinnamic acid, 2-methylcinnamic acid, predominantly trans, 2e-3-2-methylphenyl acrylic acid, e-3-2-methylphenyl-2-propenoic acid, 2e-3-2-methylphenyl prop-2-enoic acid, e-3-o-tolyl acrylic acid, e-3-o-tolylacrylic acid, 3-o-tolyl-acrylic acid
PubChem CID 819020
IUPAC Name (E)-3-(2-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=CC=C1C=CC(=O)O

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 25 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylcinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.