Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylcyclohexanone, 98%

Catalog No. AC126610010
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC126610010 1 kg
AC126610500 50 g
AC126612500 250 g
AC126610250 25 g
AC126611000 100 g
AC126615000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. AC126610010 Supplier Thermo Scientific Chemicals Supplier No. 126610010
Only null left
Add to Cart

CAS: 583-60-8 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-60-8
InChI Key LFSAPCRASZRSKS-UHFFFAOYSA-N
Synonym 2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone
PubChem CID 11419
IUPAC Name 2-methylcyclohexan-1-one
SMILES CC1CCCCC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -14.0°C
Color Colorless to Yellow
Density 0.9240g/mL
Boiling Point 162.0°C to 163.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12O
Refractive Index 1.4470 to 1.4490
Linear Formula CH3C6H9(=O)
Quantity 1 kg
Beilstein 07, 14
Specific Gravity 0.924
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Molecular Weight (g/mol) 112.17
Formula Weight 112.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexanone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-513-6
RTECSNumber GW1750000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.