Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyldibenzo[f,h]quinoxaline 98.0+%, TCI America™
SDP

Catalog No. M30051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30051G 1 g
M3005200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30051G Supplier TCI America Supplier No. M30051G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 536753-86-3
Molecular Formula C17H12N2
Molecular Weight (g/mol) 244.297
MDL Number MFCD16038624
InChI Key DCPGBPKLXYETTA-UHFFFAOYSA-N
PubChem CID 20720413
IUPAC Name 3-methylphenanthro[9,10-b]pyrazine
SMILES CC1=CN=C2C3=CC=CC=C3C4=CC=CC=C4C2=N1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Formula Weight 244.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyldibenzo[f,h]quinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.