Learn More
Selleck Chemical LLC 2-Methylenebutyrolactone S5148-25mg
SDP
Supplier:  Selleck Chemical LLC S514825MG
Encompass_Preferred
2-Methylenebutyrolactone (Tulipalin A MBL -methylene- -butyrolactone) also known as -methylene- -butyrolactone (MBL) (Tulipalin A) belongs to the class of sesquiterpene lactone family and is considered as cyclic analog of most common vinyl monomer methyl methacrylate (MMA)
Catalog No. 50-311-1903
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.