Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methylfuran

Catalog No. M46845100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M46845100ML 100 mL
M46845500ML 500 mL
M468455ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M46845100ML Supplier Sigma Aldrich Supplier No. M46845100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-methylfuran, a substituted furan, is considered as a potential alternative fuel for internal combustion engines
  • Its microwave spectrum has been analyzed
  • The synthesis of 2-mf from carbon sources under different reaction conditions have been reported.2-methylfuran may be used in the synthesis of exo-cis-1-methyl-7-oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride via diels-alder reaction with maleic anhydride

Specifications

CAS 534-22-5
Density 0.91 g/mL (at 25°C (literature))
Boiling Point 63°C to 66°C (lit.)
Molecular Formula C5H6O
Refractive Index n20/D 1.433 (literature)
Linear Formula C5H6O
MDL Number MFCD00003248
Quantity 100 mL
Synonym Silvan
Molecular Weight (g/mol) 82.1
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.