Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylheptane, 99%

Catalog No. p-7028811
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2622003 1 g
AAH2622006 5 g
AAH2622014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2622003 Supplier Thermo Scientific Chemicals Supplier No. H2622003
Only null left
Add to Cart

CAS: 592-27-8 | C8H18 | 114.232 g/mol

2-methylheptane is a gas detector and it is used in RD DIN40 respirator mask with gas filter type A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-methylheptane is a gas detector and it is used in RD DIN40 respirator mask with gas filter type A.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-27-8
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009518
InChI Key JVSWJIKNEAIKJW-UHFFFAOYSA-N
Synonym heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3
PubChem CID 11594
IUPAC Name 2-methylheptane
SMILES CCCCCC(C)C

Specifications

Density 0.698
Boiling Point 116°C
Flash Point 4°C (40°F)
Refractive Index 1.395
Linear Formula (CH3)2CHCH2CH2CH2CH2CH3
Quantity 1 g
UN Number UN1262
Beilstein 1696862
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 114.23
Percent Purity 99%
Chemical Name or Material 2-Methylheptane
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
EINECSNumber 209-747-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.