Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylhydroquinone, 99%

Catalog No. AAB212410B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB212410B 1000 g
AAB2124130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB212410B Supplier Thermo Scientific Chemicals Supplier No. B212410B
Add to Cart

CAS: 95-71-6 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-71-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002345
InChI Key CNHDIAIOKMXOLK-UHFFFAOYSA-N
Synonym methylhydroquinone, 2-methylhydroquinone, 2,5-dihydroxytoluene, p-toluhydroquinone, toluhydroquinone, toluquinol, p-toluquinol, tolylhydroquinone, p-toluhydroquinol, 2,5-toluenediol
PubChem CID 7253
IUPAC Name 2-methylbenzene-1,4-diol
SMILES CC1=CC(O)=CC=C1O

Specifications

Melting Point 127°C to 130°C
Boiling Point 284°C to 286°C
Flash Point 172°C (341°F)
Quantity 1000 g
Beilstein 2041489
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 2-Methylhydroquinone
Hazard Category H302-H317-H318-H341-H351
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c
EINECSNumber 202-443-7
RTECSNumber MX6700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.