Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylimidazo[4,5-c]pyridine, 96%

Catalog No. AAL1913106
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1913106 5 g
AAL19131MD 250 mg
AAL1913103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1913106 Supplier Thermo Scientific Chemicals Supplier No. L1913106
Add to Cart

CAS: 63604-59-1 | C7H7N3 | 133.154 g/mol

2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63604-59-1
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD03426025
InChI Key JWQVOMJORQXNME-UHFFFAOYSA-N
Synonym 2-methyl-3h-imidazo 4,5-c pyridine, 2-methyl-1h-imidazo 4,5-c pyridine, 2-methylimidazo 4,5-c pyridine, 8-methyl-3-deazapurine, 8-methyl-3,7,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene, acmc-20anq0, 2-methyl-5-azabenzimidazole, 3h-imidazo 4,5-c pyridine, 2-methyl, 1h-imidazo 4,5-c pyridine, 2-methyl
PubChem CID 373077
IUPAC Name 2-methyl-3H-imidazo[4,5-c]pyridine
SMILES CC1=NC2=C(N1)C=NC=C2

Specifications

Melting Point 176°C to 178°C
Quantity 5 g
Beilstein 3234
Solubility Information Insoluble in water.
Formula Weight 133.15
Percent Purity 99%
Chemical Name or Material 2-Methylimidazo[4,5-c]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.