Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylindole 99.0+%, TCI America™
SDP

Catalog No. M0346100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0346100G 100 g
M034625G 25 g
M0346500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0346100G Supplier TCI America Supplier No. M0346100G

Chemical Identifiers

CAS 95-20-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005616
InChI Key BHNHHSOHWZKFOX-UHFFFAOYSA-N
Synonym 2-methylindole, 1h-indole, 2-methyl, indole, 2-methyl, 2-methyl indole, unii-i7cn58827i, 2-methylindol, methylketole, methyl indole, 2-methyl-indole, pubchem7504
PubChem CID 7224
ChEBI CHEBI:49402
IUPAC Name 2-methyl-1H-indole
SMILES CC1=CC2=CC=CC=C2N1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 272°C
Quantity 100 g
Formula Weight 131.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylindole
EINECSNumber (5)-0131
RTECSNumber NM0345000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.