Learn More
Sigma Organic Chemistry 2-Methylpentane | 100G | 107-83-5 | MFCD00009406
Supplier: Sigma Organic Chemistry M65807100G
2-Methylpentane, 100G
About This Item:
Synonym(s): 'Isohexane'
Linear Formula: CH3CH2CH2CH(CH3)2
Molecular Weight: 86.18
CAS Number: 107-83-5
MDL Number: MFCD00009406
UNSPSC Code: 12352100
Purity: >=99%
Boiling Point: 62 C (lit.)
Melting Point: -154 C (lit.)
Density: 0.653 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.371 (lit.)
Storage: room temp
EINECS Number: 203-523-4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.