Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpyrazine, 99+%

Catalog No. AAA1414622
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1414622 100 g
AAA1414614 25 g
AAA1414636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1414622 Supplier Thermo Scientific Chemicals Supplier No. A1414622
Add to Cart

CAS: 109-08-0 | C5H6N2 | 94.117 g/mol

2-Methylpyrazine is used as flavoring agents in bread fresh bread, butter, chocolate, chocolate cocoa, coffee, meat roasted, nut almond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylpyrazine is used as flavoring agents in bread fresh bread, butter, chocolate, chocolate cocoa, coffee, meat roasted, nut almond.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-08-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006142
InChI Key CAWHJQAVHZEVTJ-UHFFFAOYSA-N
Synonym methylpyrazine, pyrazine, methyl, pyrazine, 2-methyl, 2-methyl-1,4-diazine, 2-methyl pyrazine, 2-methyl-pyrazine, unii-rvc6500u9c, fema no. 3309, ccris 2927, wln: t6n dnj b1
PubChem CID 7976
IUPAC Name 2-methylpyrazine
SMILES CC1=NC=CN=C1

Specifications

Melting Point -30°C
Density 1.028
Boiling Point 135°C to 136°C
Flash Point 50°C (122°F)
Odor Unpleasant
Refractive Index 1.505
Quantity 100 g
UN Number UN1993
Beilstein 105778
Solubility Information Fully miscible in water.
Formula Weight 94.12
Percent Purity ≥99%
Chemical Name or Material 2-Methylpyrazine
Show More Show Less
Hazard Category H226-H302
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-645-8
RTECSNumber UQ3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.