Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpyrimidine-5-carboxylic acid, 95%, Thermo Scientific™

Catalog No. AC454442500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454442500 250 mg
AC454440010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454442500 Supplier Thermo Scientific Chemicals Supplier No. 454442500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5194-32-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
InChI Key NMGIXZFBQPETOK-UHFFFAOYSA-N
Synonym 2-methyl-5-pyrimidinecarboxylic acid, 2-methylpyrimidine-5-carboxylicacid, 5-pyrimidinecarboxylic acid, 2-methyl, 2-methyl-5-pyrimidinecarboxylicacid, pubchem13290, acmc-209kwu, ksc269i7t, 2-methyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl, 2-methyl pyrimidine-5-carboxylic acid
PubChem CID 13670875
IUPAC Name 2-methylpyrimidine-5-carboxylic acid
SMILES CC1=NC=C(C=N1)C(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material 2-Methylpyrimidine-5-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.