Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpyrrolidine hydrochloride, 97%, Thermo Scientific™

Catalog No. AC458070010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458070010 1 g
AC458070050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458070010 Supplier Thermo Scientific Chemicals Supplier No. 458070010
Add to Cart

Chemical Identifiers

CAS 54677-53-1
Molecular Formula C5H11N·HCl
Molecular Weight (g/mol) 121.61
InChI Key JNEIFWYJFOEKIM-UHFFFAOYSA-N
Synonym 2-methylpyrrolidine hydrochloride, 2-methylpyrrolidine hcl, 2-methyl-pyrrolidine hydrochloride, pubchem11112, pubchem11115, acmc-1c01e, 2-methylpyrrolidinehydrochloride, 2-methylpyrrolidine-hydrogen chloride 1/1, pyrrolidine, 2-methyl-, hydrochloride 1:1, pyrrolidine, 2-methyl-, hydrochloride, 2r-9ci
PubChem CID 21907341
IUPAC Name 2-methylpyrrolidine;hydrochloride
SMILES CC1CCCN1.Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 121.61
Percent Purity 97%
Chemical Name or Material 2-Methylpyrrolidine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.