Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylquinoxaline 98.0+%, TCI America™
SDP

Catalog No. M058025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M058025G 25 g
M05805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M058025G Supplier TCI America Supplier No. M058025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7251-61-8
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006727
InChI Key ALHUXMDEZNLFTA-UHFFFAOYSA-N
Synonym quinoxaline, 2-methyl, 2-methyl quinoxaline, unii-03vu31mv6j, ccris 2948, methyl-quinoxaline, 2-methyl-quinoxaline, acmc-1biyr, 2-methylbenzopyrazine, 2-methyl-1,4-naphthyridine, 5-23-07-00219 beilstein handbook reference
PubChem CID 23686
IUPAC Name 2-methylquinoxaline
SMILES CC1=NC2=CC=CC=C2N=C1

Specifications

Boiling Point 245°C
Quantity 25 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylquinoxaline
RTECSNumber VD3450000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.