Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Methyltetrahydrofuran | 96-47-9 | MFCD00005367 | 1 L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4142471L
Encompass
2-Methyltetrahydrofuran | Purity: 99% | Mol Wt: 86.13 | 96-47-9 | MFCD00005367 | 1 L
Catalog No. NC1625824
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.