Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran-3-one 98.0+%, TCI America™
SDP

Catalog No. M105525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M105525G 25 g
M10555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M105525G Supplier TCI America Supplier No. M105525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3188-00-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00010423
InChI Key FCWYQRVIQDNGBI-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran-3-one, 2-methyl tetrahydro-3-furanone, 2-methyltetrahydro-3-furanone, 3 2h-furanone, dihydro-2-methyl, dihydro-2-methyl-3 2h-furanone, coffee furanone, dihydro-2-methyl-3-furanone, tetrahydro-2-methylfuran-3-one, 2-methyl-3-oxotetrahydrofuran, 2-methyl-3-ketotetrahydrofuran
PubChem CID 18522
IUPAC Name 2-methyloxolan-3-one
SMILES CC1C(=O)CCO1

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 1224
Boiling Point 139°C
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Methyltetrahydrofuran-3-one
RTECSNumber LU3579000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.