Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019125 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9125 10 mL
11-101-9977 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9125 Supplier MilliporeSigma Supelco Supplier No. 8548810MLF
Add to Cart
Add to Cart

2-Methyltetrahydrofuran is a sustainable green alternative and offers both economical and environmentally friendly advantages over tetrahydrofuran and dichloromethane in a variety of organometallic reactions and biphasic reactions.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 96-47-9
Density 0.86 g/mL (at 25°C)
Flash Point -10°C
Physical Form Neat
Linear Formula C5H10 O
MDL Number MFCD00005367
Boiling Point 78°C to 80°C
Synonym 2-MeTHF; Tetrahydro-2-methylfuran; Tetrahydrosilvan
Type Analytical Standard
Quantity 10 mL
Formula Weight 86.13
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.