Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrothiophene, 98+%, Thermo Scientific™

Catalog No. AA43456AB
Click to view available options
Quantity:
10 mL

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1795-09-1
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.20
MDL Number MFCD00015539
InChI Key AJPGNQYBSTXCJE-UHFFFAOYNA-N
Synonym 2-methyltetrahydrothiophene, thiophene, tetrahydro-2-methyl, tetrahydro-2-methylthiophene, thiophene, tetrahydromethyl, 2-methylthiophane, 2-methylthi-olane, pubchem12832, 2-methylthiacyclopentane, acmc-209efp, 2-methyl tetrahydrothiophene
PubChem CID 15711
IUPAC Name 2-methylthiolane
SMILES CC1CCCS1

Specifications

Density 0.96 g/mL
Boiling Point 132°C
Odor Stench
Quantity 10 mL
UN Number UN1993
Formula Weight 102.2
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Methyltetrahydrothiophene
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 217-270-2
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.