Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylthiazole-5-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. M26261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26261G 1 g
M26265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26261G Supplier TCI America Supplier No. M26261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40004-69-1
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD01760300
InChI Key QCXCIYPOMMIBHO-UHFFFAOYSA-N
PubChem CID 98845
IUPAC Name 2-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=NC=C(S1)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 143.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylthiazole-5-carboxylic Acid
RTECSNumber XJ3678500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.