Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)-2-thiazoline 98.0+%, TCI America™
SDP

Catalog No. M126225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M126225G 25 g
M12625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M126225G Supplier TCI America Supplier No. M126225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19975-56-5
Molecular Formula C4H7NS2
Molecular Weight (g/mol) 133.23
MDL Number MFCD00005312
InChI Key QFGRBBWYHIYNIB-UHFFFAOYSA-N
Synonym 2-(Methylmercapto)-2-thiazoline
PubChem CID 88324
IUPAC Name 2-(methylsulfanyl)-4,5-dihydro-1,3-thiazole
SMILES CSC1=NCCS1

Specifications

Color Yellow
Boiling Point 217°C
Quantity 25 g
Formula Weight 133.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)-2-thiazoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.